missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-indole-5-carboxylic acid, 95%, ACROS Organics™

Chemical Identifiers

CAS 186129-25-9
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
InChI Key UHQAIJFIXCOBCN-UHFFFAOYSA-N
Synonym 1-methyl-1h-indol-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylicacid, 1-methyl-5-indolecarboxylic acid, 1-methyl-5-indolecarboxylicacid, 1h-indole-5-carboxylic acid, 1-methyl, 1h-indole-5-carboxylicacid,1-methyl, 5-carboxy-1-methyl-1h-indole, methyl 1h-indole-5-carboxylic acid, n-methylindol-5-carboxylic acid
PubChem CID 7015601
IUPAC Name 1-methylindole-5-carboxylic acid
SMILES CN1C=CC2=C1C=CC(=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC448640010
SDS View Documents
Thermo Fisher Scientific
AC448640010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC448640050
SDS View Documents
Thermo Fisher Scientific
AC448640050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

186129-25-9
175.19
1-methyl-1h-indol-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylicacid, 1-methyl-5-indolecarboxylic acid, 1-methyl-5-indolecarboxylicacid, 1h-indole-5-carboxylic acid, 1-methyl, 1h-indole-5-carboxylicacid,1-methyl, 5-carboxy-1-methyl-1h-indole, methyl 1h-indole-5-carboxylic acid, n-methylindol-5-carboxylic acid
1-methylindole-5-carboxylic acid
C10H9NO2
UHQAIJFIXCOBCN-UHFFFAOYSA-N
7015601
CN1C=CC2=C1C=CC(=C2)C(=O)O

Specifications

Glass bottle
1g
94% min. (HPLC)
1-methyl-1h-indol-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylicacid, 1-methyl-5-indolecarboxylic acid, 1-methyl-5-indolecarboxylicacid, 1h-indole-5-carboxylic acid, 1-methyl, 1h-indole-5-carboxylicacid,1-methyl, 5-carboxy-1-methyl-1h-indole, methyl 1h-indole-5-carboxylic acid, n-methylindol-5-carboxylic acid
CN1C=CC2=C1C=CC(=C2)C(=O)O
175.19
175.19
1-Methyl-1H-indole-5-carboxylic acid
221.0°C to 223.0°C
186129-25-9
C10H9NO2
UHQAIJFIXCOBCN-UHFFFAOYSA-N
1-methylindole-5-carboxylic acid
7015601
95%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning