missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-1H-indole-5-carboxylic acid, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC448640050
Specifications
1-Methyl-1H-indole-5-carboxylic acid | |
221.0°C to 223.0°C | |
186129-25-9 | |
C10H9NO2 | |
UHQAIJFIXCOBCN-UHFFFAOYSA-N | |
1-methylindole-5-carboxylic acid | |
7015601 | |
95% |
Glass bottle | |
5g | |
94% min. (HPLC) | |
1-methyl-1h-indol-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylicacid, 1-methyl-5-indolecarboxylic acid, 1-methyl-5-indolecarboxylicacid, 1h-indole-5-carboxylic acid, 1-methyl, 1h-indole-5-carboxylicacid,1-methyl, 5-carboxy-1-methyl-1h-indole, methyl 1h-indole-5-carboxylic acid, n-methylindol-5-carboxylic acid | |
CN1C=CC2=C1C=CC(=C2)C(=O)O | |
175.19 | |
175.19 |
Chemical Identifiers
186129-25-9 | |
175.19 | |
1-methyl-1h-indol-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylic acid, 1-methyl-1h-indole-5-carboxylicacid, 1-methyl-5-indolecarboxylic acid, 1-methyl-5-indolecarboxylicacid, 1h-indole-5-carboxylic acid, 1-methyl, 1h-indole-5-carboxylicacid,1-methyl, 5-carboxy-1-methyl-1h-indole, methyl 1h-indole-5-carboxylic acid, n-methylindol-5-carboxylic acid | |
1-methylindole-5-carboxylic acid |
C10H9NO2 | |
UHQAIJFIXCOBCN-UHFFFAOYSA-N | |
7015601 | |
CN1C=CC2=C1C=CC(=C2)C(=O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning