missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-methyl-1H-indole-4-carbonitrile, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC45816CB
Specifications
1-Methyl-1H-indole-4-carbonitrile | |
97% | |
MFCD08690265 | |
1-methyl-1h-indole-4-carbonitrile, 1h-indole-4-carbonitrile,1-methyl, 1h-indole-4-carbonitrile, 1-methyl | |
CN1C=CC2=C(C=CC=C12)C#N | |
156.19 | |
156.19 |
628711-58-0 | |
C10H8N2 | |
250mg | |
DYOVHBKIEBCIIE-UHFFFAOYSA-N | |
1-methylindole-4-carbonitrile | |
22391592 | |
97% |
Chemical Identifiers
628711-58-0 | |
156.19 | |
DYOVHBKIEBCIIE-UHFFFAOYSA-N | |
22391592 | |
CN1C=CC2=C(C=CC=C12)C#N |
C10H8N2 | |
MFCD08690265 | |
1-methyl-1h-indole-4-carbonitrile, 1h-indole-4-carbonitrile,1-methyl, 1h-indole-4-carbonitrile, 1-methyl | |
1-methylindole-4-carbonitrile |
Safety and Handling
GHS Signal Word: Warning