Learn More
(1-Methyl-1H-indol-7-yl)methanol, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 854778-61-3 |
---|---|
Molecular Formula | C10H11NO |
Molecular Weight (g/mol) | 161.20 |
MDL Number | MFCD09879959 |
InChI Key | XWTPJITYDAXWKC-UHFFFAOYSA-N |
Synonym | 1-methyl-1h-indol-7-yl methanol, 1-methylindol-7-yl methanol, 1-methyl-1h-indol-7-yl-methanol, 1-methylindol-7-yl methan-1-ol |
PubChem CID | 24229741 |
IUPAC Name | (1-methyl-1H-indol-7-yl)methanol |
SMILES | CN1C=CC2=CC=CC(CO)=C12 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC66909CB
|
Thermo Scientific Chemicals
CC66909CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC66909DA
|
Thermo Scientific Chemicals
CC66909DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
854778-61-3 | |
161.20 | |
XWTPJITYDAXWKC-UHFFFAOYSA-N | |
24229741 | |
CN1C=CC2=CC=CC(CO)=C12 |
C10H11NO | |
MFCD09879959 | |
1-methyl-1h-indol-7-yl methanol, 1-methylindol-7-yl methanol, 1-methyl-1h-indol-7-yl-methanol, 1-methylindol-7-yl methan-1-ol | |
(1-methyl-1H-indol-7-yl)methanol |
Specifications
854778-61-3 | |
100.0 | |
Amber Glass Bottle | |
MFCD09879959 | |
1-methyl-1h-indol-7-yl methanol, 1-methylindol-7-yl methanol, 1-methyl-1h-indol-7-yl-methanol, 1-methylindol-7-yl methan-1-ol | |
CN1C=CC2=CC=CC(CO)=C12 | |
161.20 | |
161.2 | |
(1-Methyl-1H-indol-7-yl)methanol |
96.5 | |
97% | |
C10H11NO | |
250mg | |
XWTPJITYDAXWKC-UHFFFAOYSA-N | |
(1-methyl-1H-indol-7-yl)methanol | |
24229741 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning