Please login to your online account to display your discounted pricing

(1-Methyl-1H-indol-5-yl)methylamine, 97%, Maybridge

$402.44 - $402.44

Chemical Identifiers

Chemical Name or Material (1-Methyl-1H-indol-5-yl)methylamine
CAS 884507-17-9
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H12N2
Formula Weight 160.22
MDL Number MFCD09025839
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC41413DA

  SDS
fisher scientific company
CC41413DA
Amber Glass Bottle 1g Each for $402.44

CC41413CB

  SDS
fisher scientific company
CC41413CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.22
Quantity 1g
Assay 95%
CAS Max % 100.0
CAS 884507-17-9
CAS Min % 94.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C10H12N2
MDL Number MFCD09025839
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material (1-Methyl-1H-indol-5-yl)methylamine
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

(1-Methyl-1H-indol-5-yl)methylamine, 95%

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.