missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-5-carboxylic acid, 97%, Maybridge

Manufacturer:  Thermo Scientific CC23601DA

 View more versions of this product

Catalog No. CC23601DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Methyl-1H-imidazole-5-carboxylic acid
1g
96.5
97%
MFCD00955677
PBEDVTDUVXFSMW-UHFFFAOYSA-N
3-methylimidazole-4-carboxylic acid
2773507
97%
Amber glass bottle
41806-40-0
100.0
C5H6N2O2
1-methyl-1h-imidazole-5-carboxylic acid, 1-methylimidazole-5-carboxylic acid, 5-carboxy-1-methyl-1h-imidazole, 3-methyl-3h-imidazole-4-carboxylic acid, 1-methyl-1h-imidazole-5-carboxylicacid, 1-methyl-1h-5-imidazolecarboxylic acid, pubchem20954, acmc-209jlu, ksc236i7p, 1-methyl-5-imidazolecarboxylic acid
CN1C=NC=C1C(=O)O
126.115
126.11

Chemical Identifiers

41806-40-0
126.115
PBEDVTDUVXFSMW-UHFFFAOYSA-N
2773507
CN1C=NC=C1C(=O)O
C5H6N2O2
MFCD00955677
1-methyl-1h-imidazole-5-carboxylic acid, 1-methylimidazole-5-carboxylic acid, 5-carboxy-1-methyl-1h-imidazole, 3-methyl-3h-imidazole-4-carboxylic acid, 1-methyl-1h-imidazole-5-carboxylicacid, 1-methyl-1h-5-imidazolecarboxylic acid, pubchem20954, acmc-209jlu, ksc236i7p, 1-methyl-5-imidazolecarboxylic acid
3-methylimidazole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents