Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-5-carbonitrile, 97+%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-imidazole-5-carbonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5N3
Formula Weight 107.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC23616DE

 
fisher scientific company
CC23616DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC23616CB

 
fisher scientific company
CC23616CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 107.12
Quantity 5g
Assay 97%
Chemical Name or Material 1-Methyl-1H-imidazole-5-carbonitrile
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H5N3
CAS 66121-66-2