Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-5-carbonitrile, 97+%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC23616CB

 View more versions of this product

Catalog No. CC23616CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 107.12
Quantity 250mg
Assay 97%
CAS 66121-66-2
Packaging Amber Glass Bottle
Chemical Name or Material 1-Methyl-1H-imidazole-5-carbonitrile
Assay Percent Range 97%
Molecular Formula C5H5N3

Specifications

Chemical Name or Material 1-Methyl-1H-imidazole-5-carbonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5N3
Formula Weight 107.12