Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-4-carboxylic acid, 97%, Maybridge

$21.41 - $21.41

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6N2O2
Formula Weight 126.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC03601DA

 
fisher scientific company
CC03601DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC03601CB

 
fisher scientific company
CC03601CB
Amber Glass Bottle 250mg Each for $21.41
Description & Specifications

Specifications

Formula Weight 126.11
Quantity 1g
Assay 97%
Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid
Molecular Formula C5H6N2O2
CAS 41716-18-1
Packaging Amber Glass Bottle
Assay Percent Range 97%