Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-4-carboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC03601DA

 View more versions of this product

Catalog No. CC03601DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 126.11
Quantity 1g
Assay 97%
Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid
Molecular Formula C5H6N2O2
CAS 41716-18-1
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6N2O2
Formula Weight 126.11