Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-4-carboxylic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC03601CB

 View more versions of this product

Catalog No. CC03601CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 126.11
Quantity 250mg
Assay 97%
CAS 41716-18-1
Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid
Molecular Formula C5H6N2O2
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6N2O2
Formula Weight 126.11