Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-2-carboxylic acid, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-imidazole-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6N2O2
Formula Weight 126.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC20801EA

 
fisher scientific company
CC20801EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC20801DA

  SDS
fisher scientific company
CC20801DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC20801DE

 
fisher scientific company
CC20801DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.11
Quantity 10g
Assay 97%
Packaging Amber glass bottle
Chemical Name or Material 1-Methyl-1H-imidazole-2-carboxylic acid
Assay Percent Range 97%
Molecular Formula C5H6N2O2
CAS 20485-43-2