Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-2-carboxylic acid, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC20801DE

 View more versions of this product

Catalog No. CC20801DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 126.11
Quantity 5g
Assay 97%
CAS 20485-43-2
Assay Percent Range 97%
Molecular Formula C5H6N2O2
Chemical Name or Material 1-Methyl-1H-imidazole-2-carboxylic acid

Specifications

Chemical Name or Material 1-Methyl-1H-imidazole-2-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6N2O2
Formula Weight 126.11