missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1-Methyl-1H-imidazol-4-yl)methanol, 97%, Maybridge

Chemical Identifiers

CAS 17289-25-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H8N2O
Formula Weight 112.13
MDL Number MFCD06738905
Chemical Name or Material (1-Methyl-1H-imidazol-4-yl)methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC03609CB SDS Fisher Scientific Company
CC03609CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

17289-25-7
100.0
C5H8N2O
MFCD06738905
97%
96.5
97%
112.13
(1-Methyl-1H-imidazol-4-yl)methanol

Specifications

17289-25-7
100.0
97%
112.13
250mg
97%
96.5
Amber Glass Bottle
C5H8N2O
MFCD06738905
(1-Methyl-1H-imidazol-4-yl)methanol

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning