Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-1H-benzotriazole, 98+%

$102.05 - $343.26

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-benzotriazole
CAS 13351-73-0
Molecular Formula C7H7N3
Formula Weight 133.15
MDL Number MFCD00014572
Beilstein 118900
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0029706

 
alfa aesar™
L0029706
5g Each for $102.05

AAL0029714

 
alfa aesar™
L0029714
25g Each for $343.26
Description & Specifications

Specifications

Formula Weight 133.15
Chemical Name or Material 1-Methyl-1H-benzotriazole
Assay ≥98%
Molecular Formula C7H7N3
MDL Number MFCD00014572
CAS 13351-73-0
Beilstein 118900
Melting Point 63° to 66°C
Quantity 5g
Odor Odorless
RTECS Number DM1330000