Learn More
1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13652CB
Specifications
1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride | |
89.5 | |
90% | |
C9H8Cl2N2O | |
250 mg | |
SGNORNZSRSQMEE-UHFFFAOYSA-N | |
1-methylbenzimidazole-5-carbonyl chloride;hydrochloride | |
24229458 | |
90% |
906352-57-6 | |
100.0 | |
Amber Glass Bottle | |
MFCD09817460 | |
1-methyl-1h-benzimidazole-5-carbonyl chloride hydrochloride, 1-methyl-1,3-benzodiazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzoimidazole-5-carbonyl chloride.hcl, 1-methylbenzimidazole-5-carbonyl chloride, chloride, 1-methyl-1h-benzo d imidazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzimidazole-5-carbonyl chloride-hydrogen chloride 1/1, 1h-benzimidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1 | |
Cl.CN1C=NC2=CC(=CC=C12)C(Cl)=O | |
231.08 | |
231.08 |
Chemical Identifiers
906352-57-6 | |
231.08 | |
SGNORNZSRSQMEE-UHFFFAOYSA-N | |
24229458 | |
Cl.CN1C=NC2=CC(=CC=C12)C(Cl)=O |
C9H8Cl2N2O | |
MFCD09817460 | |
1-methyl-1h-benzimidazole-5-carbonyl chloride hydrochloride, 1-methyl-1,3-benzodiazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzoimidazole-5-carbonyl chloride.hcl, 1-methylbenzimidazole-5-carbonyl chloride, chloride, 1-methyl-1h-benzo d imidazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzimidazole-5-carbonyl chloride-hydrogen chloride 1/1, 1h-benzimidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1 | |
1-methylbenzimidazole-5-carbonyl chloride;hydrochloride |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger