Please login to your online account to display your discounted pricing

(1-Methyl-1H-benzimidazol-2-yl)methylamine, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC03813DE

 View more versions of this product

Catalog No. CC03813DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.21
Quantity 5g
Assay 97%
Chemical Name or Material (1-Methyl-1H-benzimidazol-2-yl)methylamine
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H11N3
CAS 20028-40-4

Specifications

Chemical Name or Material (1-Methyl-1H-benzimidazol-2-yl)methylamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H11N3
Formula Weight 161.21