Please login to your online account to display your discounted pricing

(1-Methyl-1H-benzimidazol-2-yl)methylamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material (1-Methyl-1H-benzimidazol-2-yl)methylamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H11N3
Formula Weight 161.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC03813DE

 
fisher scientific company
CC03813DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.21
Quantity 5g
Assay 97%
Chemical Name or Material (1-Methyl-1H-benzimidazol-2-yl)methylamine
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H11N3
CAS 20028-40-4