Please login to your online account to display your discounted pricing

1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC23504DE

 View more versions of this product

Catalog No. CC23504DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.16
Quantity 5g
Assay 97%
Assay Percent Range 97%
Packaging Amber Glass Bottle
CAS 499770-67-1
Molecular Formula C8H7N3O
Chemical Name or Material 1-Methyl-1H-1, 2, 3-benzotriazole-5-carbaldehyde

Specifications

Chemical Name or Material 1-Methyl-1H-1, 2, 3-benzotriazole-5-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7N3O
Formula Weight 161.16