Please login to your online account to display your discounted pricing

1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Methyl-1-propenylmagnesium bromide
CAS 85676-85-3
CAS Min % 7.68
CAS Max % 9.38
CAS 109-99-9
CAS Min % 90.62
CAS Max % 92.32
Molecular Formula C4H7BrMg
Formula Weight 159.31
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC436021000

  SDS
thermo fisher scientific
AC436021000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC436028000

  SDS
thermo fisher scientific
AC436028000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Store in a dry place. Store in a closed container. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
May cause respiratory irritation. Causes severe skin burns and eye damage. Highly flammable liquid and vapor. In contact with water releases flammable gases. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.
Formula Weight 159.31
Physical Form Liquid
Quantity 100mL
Color Colorless to dark yellow
Packaging AcroSeal™ Glass Bottle
Density 0.9340g/mL
Molecular Formula C4H7BrMg
CAS 85676-85-3
CAS Min % 90.62
CAS Max % 92.32
CAS 109-99-9
CAS Min % 7.68
CAS Max % 9.38
Chemical Name or Material 1-Methyl-1-propenylmagnesium bromide
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Store in a dry place. Store in a closed container. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
May cause respiratory irritation. Causes severe skin burns and eye damage. Highly flammable liquid and vapor. In contact with water releases flammable gases. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.