missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1-phenylhydrazine, TCI America™

Manufacturer:  TCI America M039925G

 View more versions of this product

Catalog No. M039925G



Specifications

Specifications

1-Methyl-1-phenylhydrazine
Undesignated
618-40-6
MFCD00007627
MWOODERJGVWYJE-UHFFFAOYSA-N
1-methyl-1-phenylhydrazine
36299
Clear Liquid at 20°C
109°C
25g
C7H10N2
1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
CN(C1=CC=CC=C1)N
122.171
122.17
≥95.0% (GC)

Chemical Identifiers

618-40-6
122.171
MWOODERJGVWYJE-UHFFFAOYSA-N
36299
CN(C1=CC=CC=C1)N
C7H10N2
MFCD00007627
1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
1-methyl-1-phenylhydrazine
Safety and Handling

Safety and Handling

RTECSNumber : MV7712000

TSCA : Yes

Documents