missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-1-p-tolylethylamine, 97%

Manufacturer:  Alfa Aesar™ H6623203

 View more versions of this product

Catalog No. AAH6623203



Specifications

Specifications

1-Methyl-1-p-tolylethylamine
6526-79-0
MFCD08669637
2-4-methylphenyl propan-2-amine, 2-p-tolylpropan-2-amine, 1-4-methylphenyl-1-methylethylamine, 1-methyl-1-p-tolylethylamine, 2-p-tolyl propan-2-amine, 2-4-methylphenyl-2-propanamine, 2-4-methylphenyl prop-2-ylamine, 1-methyl-1-p-tolyl-ethylamine, benzenemethanamine, alpha,alpha,4-trimethyl, alpha-amino-4-methylcumene
CC1=CC=C(C=C1)C(C)(C)N
149.237
149.12
97%
1g
C10H15N
2637596
AHIOEYFAAFXSSZ-UHFFFAOYSA-N
2-(4-methylphenyl)propan-2-amine
584050
Solid

Chemical Identifiers

6526-79-0
149.237
AHIOEYFAAFXSSZ-UHFFFAOYSA-N
584050
CC1=CC=C(C=C1)C(C)(C)N
C10H15N
MFCD08669637
2-4-methylphenyl propan-2-amine, 2-p-tolylpropan-2-amine, 1-4-methylphenyl-1-methylethylamine, 1-methyl-1-p-tolylethylamine, 2-p-tolyl propan-2-amine, 2-4-methylphenyl-2-propanamine, 2-4-methylphenyl prop-2-ylamine, 1-methyl-1-p-tolyl-ethylamine, benzenemethanamine, alpha,alpha,4-trimethyl, alpha-amino-4-methylcumene
2-(4-methylphenyl)propan-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents