missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1-(m-tolyl)hydrazine, TCI America™

$334.59 - $869.33

Chemical Identifiers

CAS 72233-91-1
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD03844802
InChI Key ABANUKHECQXXRH-UHFFFAOYSA-N
PubChem CID 827964
IUPAC Name 1-methyl-1-(3-methylphenyl)hydrazine
SMILES CC1=CC(=CC=C1)N(C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14195G View Documents TCI America
M14195G
5g Each for $334.59
 
M141925G View Documents TCI America
M141925G
25g Each for $869.33
 
Specifications

Chemical Identifiers

72233-91-1
136.198
ABANUKHECQXXRH-UHFFFAOYSA-N
1-methyl-1-(3-methylphenyl)hydrazine
C8H12N2
MFCD03844802
827964
CC1=CC(=CC=C1)N(C)N

Specifications

Undesignated
72233-91-1
MFCD03844802
CC1=CC(=CC=C1)N(C)N
136.198
136.20
≥97.0% (T)
5g
C8H12N2
ABANUKHECQXXRH-UHFFFAOYSA-N
1-methyl-1-(3-methylphenyl)hydrazine
827964
Clear Liquid at 20°C
1-Methyl-1-(m-tolyl)hydrazine
Safety and Handling

Safety and Handling

TSCA : No