missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1-cyclopentene 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 126700050

 View more versions of this product

Catalog No. AC126700050


 

Specifications Description & Specifications

Specifications

1-Methyl-1-cyclopentene
Clear colorless
96%
95.0
Authentic
1.4310 to 1.4330 (20°C, 589nm)
≥95%
C6H10
82.13
05, 64
0.78g/mL
5mL
693-89-0
100.0
Glass bottle
Liquid
96%
C5H7CH3
MFCD00001397

Chemical Identifiers

693-89-0
100.0
C6H10
82.13
1-Methyl-1-cyclopentene, 96%
95.0
96%
C5H7CH3
MFCD00001397
≥95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-762-0

TSCA : TSCA