Please login to your online account to display your discounted pricing

1-Methyl-1-cyclopentene 96%, ACROS Organics™

$87.65 - $244.25

Chemical Identifiers

Chemical Name or Material 1-Methyl-1-cyclopentene
Name Note 96%
CAS 693-89-0
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C6H10
Linear Formula C5H7CH3
Formula Weight 82.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126700050

  SDS
thermo fisher scientific
126700050
Glass bottle 5mL Each for $87.65

AC126700250

  SDS
thermo fisher scientific
126700250
Glass bottle 25mL Each for $244.25
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 82.13
Quantity 5mL
Color Clear colorless
Beilstein 05, 64
EINECS Number 211-762-0
TSCA TSCA
Density 0.78g/mL
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 1-Methyl-1-cyclopentene
Refractive Index 1.4310 to 1.4330 (20°C, 589nm)
Name Note 96%
Assay Percent Range 96%
Molecular Formula C6H10
Linear Formula C5H7CH3
MDL Number MFCD00001397
CAS 693-89-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.