missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1,2,3,4-tetrahydroisoquinoline, TCI America™

$128.34 - $378.89

Chemical Identifiers

CAS 4965-09-7
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00798989
InChI Key QPILYVQSKNWRDD-UHFFFAOYNA-N
Synonym 1-m-thiq, 1-methyl-1,2,3,4-tetrahydro-isoquinoline, 1,2,3,4-tetrahydro-1-methylisoquinoline, chembl95456, isoquinoline, tetrahydromethyl, n-methyl,tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro-1-methyl, 1metiq, pubchem13018, acmc-1aktb
PubChem CID 92214
IUPAC Name 1-methyl-1,2,3,4-tetrahydroisoquinoline
SMILES CC1C2=CC=CC=C2CCN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26065G View Documents TCI America
M26065G
5g Each for $378.89
 
M26061G View Documents TCI America
M26061G
1g Each for $128.34
 
Specifications

Chemical Identifiers

4965-09-7
147.22
QPILYVQSKNWRDD-UHFFFAOYNA-N
92214
CC1C2=CC=CC=C2CCN1
C10H13N
MFCD00798989
1-m-thiq, 1-methyl-1,2,3,4-tetrahydro-isoquinoline, 1,2,3,4-tetrahydro-1-methylisoquinoline, chembl95456, isoquinoline, tetrahydromethyl, n-methyl,tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro-1-methyl, 1metiq, pubchem13018, acmc-1aktb
1-methyl-1,2,3,4-tetrahydroisoquinoline

Specifications

228°C
5g
C10H13N
1-m-thiq, 1-methyl-1,2,3,4-tetrahydro-isoquinoline, 1,2,3,4-tetrahydro-1-methylisoquinoline, chembl95456, isoquinoline, tetrahydromethyl, n-methyl,tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro-1-methyl, 1metiq, pubchem13018, acmc-1aktb
CC1C2=CC=CC=C2CCN1
147.22
147.22
≥98.0% (T)
Yellow
4965-09-7
MFCD00798989
QPILYVQSKNWRDD-UHFFFAOYNA-N
1-methyl-1,2,3,4-tetrahydroisoquinoline
92214
Clear Liquid at 20°C
1-Methyl-1,2,3,4-tetrahydroisoquinoline
Safety and Handling

Safety and Handling

RTECSNumber : NX5087850

TSCA : No

Recommended Storage : Refrigerator