missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-1,2,3,4-tetrahydroisoquinoline, 95%

Manufacturer:  Alfa Aesar™ H6419103

 View more versions of this product

Catalog No. AAH6419103



Description

Description

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

1-Methyl-1,2,3,4-tetrahydroisoquinoline
Yellow
4965-09-7
MFCD00798989
1-m-thiq, 1-methyl-1,2,3,4-tetrahydro-isoquinoline, 1,2,3,4-tetrahydro-1-methylisoquinoline, chembl95456, isoquinoline, tetrahydromethyl, n-methyl,tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro-1-methyl, 1metiq, pubchem13018, acmc-1aktb
CC1C2=CC=CC=C2CCN1
147.22
147.22
95%
119°C (18mmHg)
1g
C10H13N
82140
QPILYVQSKNWRDD-UHFFFAOYNA-N
1-methyl-1,2,3,4-tetrahydroisoquinoline
92214
Liquid

Chemical Identifiers

4965-09-7
147.22
QPILYVQSKNWRDD-UHFFFAOYNA-N
92214
CC1C2=CC=CC=C2CCN1
C10H13N
MFCD00798989
1-m-thiq, 1-methyl-1,2,3,4-tetrahydro-isoquinoline, 1,2,3,4-tetrahydro-1-methylisoquinoline, chembl95456, isoquinoline, tetrahydromethyl, n-methyl,tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro-1-methyl, 1metiq, pubchem13018, acmc-1aktb
1-methyl-1,2,3,4-tetrahydroisoquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : NX5087850

storageNote1 : -30° to -10°C

SDS Documents