missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-1,2,3,4-tetrahydroisoquinoline, 95%

Chemical Identifiers

CAS 4965-09-7
Formula Weight 147.22
MDL Number MFCD00798989
Beilstein 82140
Chemical Name or Material 1-Methyl-1,2,3,4-tetrahydroisoquinoline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6419103 SDS Alfa Aesar™
H6419103
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6419106 SDS Alfa Aesar™
H6419106
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

4965-09-7
MFCD00798989
1-Methyl-1,2,3,4-tetrahydroisoquinoline
147.22
82140

Specifications

119°C (18mmHg)
1g
Liquid
MFCD00798989
1-Methyl-1,2,3,4-tetrahydroisoquinoline
Pale yellow
4965-09-7
147.22
82140
95%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : NX5087850

storageNote1 : -30° to -10°C