missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Methoxymethyl)-1H-benzotriazole, TCI America™

$129.86 - $385.02

Chemical Identifiers

CAS 71878-80-3
Molecular Formula C8H9N3O
Molecular Weight (g/mol) 163.18
MDL Number MFCD00216630
InChI Key QEIXBXXKTUNWDK-UHFFFAOYSA-N
PubChem CID 542390
IUPAC Name 1-(methoxymethyl)benzotriazole
SMILES COCN1C2=CC=CC=C2N=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12761G View Documents TCI America
M12761G
1g Each for $129.86
 
M12765G View Documents TCI America
M12765G
5g Each for $385.02
 
Specifications

Chemical Identifiers

71878-80-3
163.18
QEIXBXXKTUNWDK-UHFFFAOYSA-N
1-(methoxymethyl)benzotriazole
C8H9N3O
MFCD00216630
542390
COCN1C2=CC=CC=C2N=N1

Specifications

White-Yellow
1g
C8H9N3O
QEIXBXXKTUNWDK-UHFFFAOYSA-N
1-(methoxymethyl)benzotriazole
542390
Crystal-Powder at 20°C
1-(Methoxymethyl)-1H-benzotriazole
40°C
71878-80-3
MFCD00216630
COCN1C2=CC=CC=C2N=N1
163.18
163.18
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No