missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene 95.0+%, TCI America™

Manufacturer:  TCI America M075925ML

 View more versions of this product

Catalog No. M075925ML



Specifications

Specifications

1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene
25mL
C8H16O2Si
1993
SHALBPKEGDBVKK-SREVYHEPSA-N
[(3Z)-4-methoxybuta-1,3-dien-2-yl]oxy-trimethylsilane
6436851
Clear Liquid at 20°C
Colorless
59414-23-2
MFCD00008498
1-methoxy-3-trimethylsiloxy-1,3-butadiene
COC=CC(=C)O[Si](C)(C)C
172.299
172.30
≥95.0% (GC)

Chemical Identifiers

59414-23-2
172.299
SHALBPKEGDBVKK-SREVYHEPSA-N
6436851
COC=CC(=C)O[Si](C)(C)C
C8H16O2Si
MFCD00008498
1-methoxy-3-trimethylsiloxy-1,3-butadiene
[(3Z)-4-methoxybuta-1,3-dien-2-yl]oxy-trimethylsilane
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

Documents