missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methoxy-2-butanol, TCI America™

$64.17 - $436.95

Chemical Identifiers

CAS 53778-73-7
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00059143
InChI Key CSZZMFWKAQEMPB-UHFFFAOYSA-N
Synonym 1,2-Butanediol 1-Monomethyl Ether
PubChem CID 40895
IUPAC Name 1-methoxybutan-2-ol
SMILES CCC(COC)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M074725ML View Documents TCI America
M074725ML
25mL Each for $64.17
 
M0747500ML View Documents TCI America
M0747500ML
500mL Each for $436.95
 
Specifications

Chemical Identifiers

53778-73-7
104.149
CSZZMFWKAQEMPB-UHFFFAOYSA-N
40895
CCC(COC)O
C5H12O2
MFCD00059143
1,2-Butanediol 1-Monomethyl Ether
1-methoxybutan-2-ol

Specifications

135°C
25mL
C5H12O2
1993
CSZZMFWKAQEMPB-UHFFFAOYSA-N
1-methoxybutan-2-ol
40895
Clear Liquid at 20°C
1-Methoxy-2-butanol
Yellow
53778-73-7
MFCD00059143
1,2-Butanediol 1-Monomethyl Ether
CCC(COC)O
104.149
104.15
≥93.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : EL4540000

TSCA : Yes