missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methoxy-1-trimethylsilyloxypropene, TCI America™

$255.14 - $880.02

Chemical Identifiers

CAS 34880-70-1
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00671554
InChI Key RTHAKVSHSCPUGI-VOTSOKGWSA-N
Synonym [(1-Methoxy-1-propenyl)oxy]trimethylsilane
PubChem CID 10855775
IUPAC Name [(E)-1-methoxyprop-1-enoxy]-trimethylsilane
SMILES CC=C(OC)O[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11991G View Documents TCI America
M11991G
1g Each for $255.14
 
M11995G View Documents TCI America
M11995G
5g Each for $880.02
 
Specifications

Chemical Identifiers

34880-70-1
160.288
RTHAKVSHSCPUGI-VOTSOKGWSA-N
10855775
CC=C(OC)O[Si](C)(C)C
C7H16O2Si
MFCD00671554
[(1-Methoxy-1-propenyl)oxy]trimethylsilane
[(E)-1-methoxyprop-1-enoxy]-trimethylsilane

Specifications

52°C
1g
C7H16O2Si
1993
RTHAKVSHSCPUGI-VOTSOKGWSA-N
[(E)-1-methoxyprop-1-enoxy]-trimethylsilane
10855775
Clear Liquid at 20°C
1-Methoxy-1-trimethylsilyloxypropene
Colorless
34880-70-1
MFCD00671554
[(1-Methoxy-1-propenyl)oxy]trimethylsilane
CC=C(OC)O[Si](C)(C)C
160.288
160.29
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator