missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Mesitylenesulfonyl)imidazole, 98+%

$47.35 - $673.91

Chemical Identifiers

CAS 50257-39-1
Molecular Formula C12H14N2O2S
Molecular Weight (g/mol) 250.316
MDL Number MFCD00005284
InChI Key XFHVQSPNDDOABS-UHFFFAOYSA-N
Synonym 1-2,4,6-trimethylbenzenesulfonyl imidazole
PubChem CID 99556
IUPAC Name 1-(2,4,6-trimethylphenyl)sulfonylimidazole
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)N2C=CN=C2)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1505306 View Documents Alfa Aesar™
A1505306
5g Each for $47.35
 
AAA1505314 View Documents Alfa Aesar™
A1505314
25g Each for $192.77
 
AAA1505322 View Documents Alfa Aesar™
A1505322
100g Each for $673.91
 
Specifications

Chemical Identifiers

50257-39-1
250.316
XFHVQSPNDDOABS-UHFFFAOYSA-N
99556
CC1=CC(=C(C(=C1)C)S(=O)(=O)N2C=CN=C2)C
C12H14N2O2S
MFCD00005284
1-2,4,6-trimethylbenzenesulfonyl imidazole
1-(2,4,6-trimethylphenyl)sulfonylimidazole

Specifications

97°C to 99°C
50257-39-1
MFCD00005284
XFHVQSPNDDOABS-UHFFFAOYSA-N
1-(2,4,6-trimethylphenyl)sulfonylimidazole
99556
≥98%
5g
C12H14N2O2S
1-2,4,6-trimethylbenzenesulfonyl imidazole
CC1=CC(=C(C(=C1)C)S(=O)(=O)N2C=CN=C2)C
250.316
250.32
1-(Mesitylenesulfonyl)imidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning