missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Mesitylenesulfonyl)imidazole, 98+%

Manufacturer:  Alfa Aesar™ A1505306

 View more versions of this product

Catalog No. AAA1505306



Specifications

Specifications

1-(Mesitylenesulfonyl)imidazole
5g
C12H14N2O2S
1-mesitylsulfonyl-1h-imidazole, 1-2-mesitylenesulfonyl imidazole, n-mesitylenesulfonyl imidazole, n-mesitylenesulfonylimidazole, 1-2,4,6-trimethylbenzenesulfonyl imidazole, 1-2,4,6-trimethylphenyl sulphonyl-1h-imidazole, 1h-imidazole, 1-2,4,6-trimethylphenyl sulfonyl, 2,4,6-trimethylphenyl sulfonyl imidazole, acmc-209kkw, mesitylenesulfonylimidazole
CC1=CC(=C(C(=C1)C)S(=O)(=O)N2C=CN=C2)C
250.316
250.32
97°C to 99°C
50257-39-1
MFCD00005284
XFHVQSPNDDOABS-UHFFFAOYSA-N
1-(2,4,6-trimethylphenyl)sulfonylimidazole
99556
≥98%

Chemical Identifiers

50257-39-1
250.316
XFHVQSPNDDOABS-UHFFFAOYSA-N
99556
CC1=CC(=C(C(=C1)C)S(=O)(=O)N2C=CN=C2)C
C12H14N2O2S
MFCD00005284
1-mesitylsulfonyl-1h-imidazole, 1-2-mesitylenesulfonyl imidazole, n-mesitylenesulfonyl imidazole, n-mesitylenesulfonylimidazole, 1-2,4,6-trimethylbenzenesulfonyl imidazole, 1-2,4,6-trimethylphenyl sulphonyl-1h-imidazole, 1h-imidazole, 1-2,4,6-trimethylphenyl sulfonyl, 2,4,6-trimethylphenyl sulfonyl imidazole, acmc-209kkw, mesitylenesulfonylimidazole
1-(2,4,6-trimethylphenyl)sulfonylimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents