missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Mercaptomethyl)cyclopropaneacetic Acid, TCI America™

$134.45 - $400.28

Chemical Identifiers

CAS 162515-68-6
Molecular Formula C6H10O2S
Molecular Weight (g/mol) 146.204
MDL Number MFCD03411600
InChI Key VFAXPOVKNPTBTM-UHFFFAOYSA-N
Synonym 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
PubChem CID 9793825
IUPAC Name 2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
SMILES C1CC1(CC(=O)O)CS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20745G View Documents TCI America
M20745G
5g Each for $400.28
 
M20741G View Documents TCI America
M20741G
1g Each for $134.45
 
Specifications

Chemical Identifiers

162515-68-6
146.204
VFAXPOVKNPTBTM-UHFFFAOYSA-N
9793825
C1CC1(CC(=O)O)CS
C6H10O2S
MFCD03411600
2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
2-[1-(sulfanylmethyl)cyclopropyl]acetic acid

Specifications

White-Yellow
5g
C6H10O2S
3261
VFAXPOVKNPTBTM-UHFFFAOYSA-N
2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
9793825
Crystal-Powder at 20°C
1-(Mercaptomethyl)cyclopropaneacetic Acid
45°C
162515-68-6
MFCD03411600
2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
C1CC1(CC(=O)O)CS
146.204
146.20
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No