missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Mercaptomethyl)cyclopropaneacetic acid, 98%

Manufacturer:  Alfa Aesar™ H6172306

 View more versions of this product

Catalog No. AAH6172306



Specifications

Specifications

1-(Mercaptomethyl)cyclopropaneacetic acid
White
5g
C6H10O2S
UN3261
VFAXPOVKNPTBTM-UHFFFAOYSA-N
2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
9793825
Crystalline
Stench
42°C to 46°C
162515-68-6
MFCD03411600
2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
C1CC1(CC(=O)O)CS
146.204
146.21
98%

Chemical Identifiers

162515-68-6
146.204
VFAXPOVKNPTBTM-UHFFFAOYSA-N
9793825
C1CC1(CC(=O)O)CS
C6H10O2S
MFCD03411600
2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H317
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents