Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Mercaptoadamantane, 98%

$40.07 - $40.07

Chemical Identifiers

Chemical Name or Material 1-Mercaptoadamantane
CAS 34301-54-7
Molecular Formula C10H16S
Formula Weight 168.3
MDL Number MFCD00213530
UN Number UN2811
Synonym Adamantane-1-thiol; 1-Adamantyl hydrosulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58958MD

  SDS
alfa aesar™
H58958MD
250mg Each for $40.07
Description & Specifications

Specifications

Formula Weight 168.3
Quantity 250mg
Chemical Name or Material 1-Mercaptoadamantane
CAS 34301-54-7
Melting Point 99° to 106°C
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Synonym Adamantane-1-thiol; 1-Adamantyl hydrosulfide
MDL Number MFCD00213530
Assay 98%
Health Hazard 2 GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Molecular Formula C10H16S
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.