missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(m-Tolyl)piperazine 95.0+%, TCI America™
$54.75 - $137.70
Chemical Identifiers
CAS | 41186-03-2 |
---|---|
Molecular Formula | C11H16N2 |
Molecular Weight (g/mol) | 176.263 |
InChI Key | JIWHIRLNKIUYSM-UHFFFAOYSA-N |
Synonym | 1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine |
PubChem CID | 83111 |
IUPAC Name | 1-(3-methylphenyl)piperazine |
SMILES | CC1=CC(=CC=C1)N2CCNCC2 |
Chemical Identifiers
41186-03-2 | |
176.263 | |
1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine | |
1-(3-methylphenyl)piperazine |
C11H16N2 | |
JIWHIRLNKIUYSM-UHFFFAOYSA-N | |
83111 | |
CC1=CC(=CC=C1)N2CCNCC2 |
Specifications
41186-03-2 | |
C11H16N2 | |
3267 | |
JIWHIRLNKIUYSM-UHFFFAOYSA-N | |
1-(3-methylphenyl)piperazine | |
83111 | |
≥95.0% (GC) | |
1-(m-Tolyl)piperazine |
156°C | |
5 g | |
1-3-methylphenyl piperazine, 1-m-tolyl piperazine, 1-m-tolylpiperazine, 1-3-methylphenyl-piperazine, 1-m-tolyl-piperazine, piperazine, 1-3-methylphenyl, 3-methylphenyl piperazine, 4-m-tolylpiperazine, 1-m-tolyl-piperzine, 4-m-tolyl-piperazine | |
CC1=CC(=CC=C1)N2CCNCC2 | |
176.263 | |
176.26 | |
Liquid |
Safety and Handling
TSCA : No