missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Isopropyl-4-piperidone 97.0+%, TCI America™

$37.86 - $105.34

Chemical Identifiers

CAS 5355-68-0
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00038035
InChI Key CCDBCHAQIXKJCG-UHFFFAOYSA-N
Synonym 1-1'-methylethyl-4-piperidone
PubChem CID 79313
IUPAC Name 1-propan-2-ylpiperidin-4-one
SMILES CC(C)N1CCC(=O)CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05665G View Documents Tci America
I05665G
5g Each for $37.86
 
I056625G View Documents Tci America
I056625G
25g Each for $105.34
 
Specifications

Chemical Identifiers

5355-68-0
141.214
CCDBCHAQIXKJCG-UHFFFAOYSA-N
79313
CC(C)N1CCC(=O)CC1
C8H15NO
MFCD00038035
1-1'-methylethyl-4-piperidone
1-propan-2-ylpiperidin-4-one

Specifications

101°C
5g
C8H15NO
1-1'-methylethyl-4-piperidone
CC(C)N1CCC(=O)CC1
141.214
141.21
≥97.0% (T)
Undesignated
5355-68-0
MFCD00038035
CCDBCHAQIXKJCG-UHFFFAOYSA-N
1-propan-2-ylpiperidin-4-one
79313
Clear Liquid at 20°C
1-Isopropyl-4-piperidone
Safety and Handling

Safety and Handling

TSCA : No