Please login to your online account to display your discounted pricing

1-Isopropyl-1-H-pyrazole-4-boronic acid pinacol ester, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 1-Isopropyl-1-H-pyrazole-4-boronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C12H21BN2O2
Formula Weight 236.12
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC445090050

  SDS
thermo fisher scientific
AC445090050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC445090010

  SDS
thermo fisher scientific
AC445090010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.12
Quantity 5g
Assay Percent Range 97%
Chemical Name or Material 1-Isopropyl-1-H-pyrazole-4-boronic acid pinacol ester
Molecular Formula C12H21BN2O2
CAS 879487-10-2