missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Isochromanone, TCI America™

$50.42 - $151.25

Chemical Identifiers

CAS 4702-34-5
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00799232
InChI Key XVTAQSGZOGYIEY-UHFFFAOYSA-N
Synonym 3,4-Dihydro-1H-2-benzopyran-1-one
PubChem CID 78429
ChEBI CHEBI:23745
IUPAC Name 3,4-dihydroisochromen-1-one
SMILES C1COC(=O)C2=CC=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09535G View Documents TCI America
I09535G
5g Each for $151.25
 
I09531G View Documents TCI America
I09531G
1g Each for $50.42
 
Specifications

Chemical Identifiers

4702-34-5
148.161
XVTAQSGZOGYIEY-UHFFFAOYSA-N
78429
3,4-dihydroisochromen-1-one
C9H8O2
MFCD00799232
3,4-Dihydro-1H-2-benzopyran-1-one
CHEBI:23745
C1COC(=O)C2=CC=CC=C21

Specifications

160°C
5g
C9H8O2
3,4-Dihydro-1H-2-benzopyran-1-one
C1COC(=O)C2=CC=CC=C21
148.161
CHEBI:23745
Clear Liquid at 20°C
1-Isochromanone
Yellow
4702-34-5
MFCD00799232
XVTAQSGZOGYIEY-UHFFFAOYSA-N
3,4-dihydroisochromen-1-one
78429
148.16
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No