missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Iodopyrene, TCI America™

$195.56 - $676.81

Chemical Identifiers

CAS 34244-15-0
Molecular Formula C16H9I
Molecular Weight (g/mol) 328.152
InChI Key CWNJSSNWLUIMDP-UHFFFAOYSA-N
PubChem CID 13555714
IUPAC Name 1-iodopyrene
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09771G View Documents TCI America
I09771G
1g Each for $676.81
 
I0977200MG View Documents TCI America
I0977200MG
200mg Each for $195.56
 
Specifications

Chemical Identifiers

34244-15-0
328.152
13555714
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)I
C16H9I
CWNJSSNWLUIMDP-UHFFFAOYSA-N
1-iodopyrene

Specifications

Yellow
1g
C16H9I
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)I
328.152
328.15
≥98.0% (GC)
91°C
34244-15-0
CWNJSSNWLUIMDP-UHFFFAOYSA-N
1-iodopyrene
13555714
Crystal-Powder at 20°C
1-Iodopyrene
Safety and Handling

Safety and Handling

TSCA : No