Please login to your online account to display your discounted pricing

1-Iodopropane, 99%, stabilized, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  174050050

 View more versions of this product

Catalog No. AC174050050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5030 to 1.5060 (20°C, 589nm)
Stabilizer copper
Density 1.7400g/mL
Color Clear colorless to yellow
Quantity 5mL
Chemical Name or Material 1-Iodopropane
Name Note 99%
CAS 107-08-4
CAS Min % 98.5
CAS Max % 100.0
Physical Form Clear colorless to yellow liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C3H7I
Linear Formula CH3CH2CH2I
Formula Weight 169.99g/mol
MDL Number MFCD00001095
Beilstein 01, 113
Merck Index 15, 7974
Synonym Propyl iodide
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number TZ4100000
EINECS Number 203-460-2
TSCA TSCA

Specifications

Chemical Name or Material 1-Iodopropane
Name Note 99%
CAS 107-08-4
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C3H7I
Linear Formula CH3CH2CH2I
Formula Weight 169.99g/mol
MDL Number MFCD00001095
Synonym Propyl iodide
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.
Harmful if inhaled.

SDS & Safety Documents