missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodoperfluoro(4-methyl-2-pentene), 94%

Manufacturer:  Alfa Aesar™ L1679806

 View more versions of this product

Catalog No. AAL1679806


 

Specifications Description & Specifications

Specifications

1-Iodoperfluoro(4-methyl-2-pentene)
94%
407.95
3618405
1-Iodo-4-(trifluoromethyl)octafluoro-2-pentene; Perfluoro(4-methyl-2-pentenyl) iodide
120695-78-5
(CF3)2CFCF=CFCF2I
MFCD00153199
32° to 33°C (50mmHg)
5g

Chemical Identifiers

120695-78-5
407.95
3618405
1-Iodoperfluoro(4-methyl-2-pentene)
(CF3)2CFCF=CFCF2I
MFCD00153199
1-Iodo-4-(trifluoromethyl)octafluoro-2-pentene; Perfluoro(4-methyl-2-pentenyl) iodide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning