Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodoperfluoro(4-methyl-2-pentene), 94%

$87.84 - $87.84

Chemical Identifiers

Chemical Name or Material 1-Iodoperfluoro(4-methyl-2-pentene)
CAS 120695-78-5
Molecular Formula (CF3)2CFCF=CFCF2I
Formula Weight 407.95
MDL Number MFCD00153199
Beilstein 3618405
Synonym 1-Iodo-4-(trifluoromethyl)octafluoro-2-pentene; Perfluoro(4-methyl-2-pentenyl) iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1679806

  SDS
alfa aesar™
L1679806
5g Each for $87.84
Description & Specifications

Specifications

Formula Weight 407.95
Chemical Name or Material 1-Iodoperfluoro(4-methyl-2-pentene)
Molecular Formula (CF3)2CFCF=CFCF2I
MDL Number MFCD00153199
CAS 120695-78-5
Beilstein 3618405
Synonym 1-Iodo-4-(trifluoromethyl)octafluoro-2-pentene; Perfluoro(4-methyl-2-pentenyl) iodide
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 94%
Boiling Point 32° to 33°C (50mmHg)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.