missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Iodopentane, 97%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 412471000

 View more versions of this product

Catalog No. AC412471000


 

Specifications Description & Specifications

Specifications

1-Iodopentane
628-17-1
100.0
Glass bottle
Liquid
97%
1.5100g/mL
CH3(CH2)4I
MFCD00001100
Clear colorless to yellow
100g
97%
96.0
Authentic
1.4940 to 1.4960 (20°C, 589nm)
≥96.0%
copper
C5H11I
198.05
01, 133
n-Amyl iodide

Chemical Identifiers

628-17-1
100.0
C5H11I
198.05
n-Amyl iodide
≥96.0%
96.0
97%
CH3(CH2)4I
MFCD00001100
1-Iodopentane, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-030-0

RTECSNumber : SA2975600

TSCA : TSCA