missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Iodopentane, 97%, stabilized, ACROS Organics™

$58.73 - $58.73

Chemical Identifiers

CAS 628-17-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H11I
Linear Formula CH3(CH2)4I
Formula Weight 198.05
MDL Number MFCD00001100
Synonym n-Amyl iodide
Chemical Name or Material 1-Iodopentane, 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC412471000 SDS Thermo Fisher Scientific
412471000
Glass bottle 100g Each for $58.73
Specifications Description & Specifications

Chemical Identifiers

628-17-1
100.0
C5H11I
198.05
n-Amyl iodide
≥96.0%
96.0
97%
CH3(CH2)4I
MFCD00001100
1-Iodopentane, 97%

Specifications

628-17-1
100.0
Glass bottle
Liquid
copper
C5H11I
198.05
01, 133
n-Amyl iodide
1-Iodopentane, 97%
96.0
Authentic
1.4940 to 1.4960 (20°C, 589nm)
97%
1.5100g/mL
CH3(CH2)4I
MFCD00001100
Clear colorless to yellow
100g
≥96.0%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-030-0

RTECSNumber : SA2975600

TSCA : TSCA