missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
1-Iodooctadecane, TCI America™
Chemical Identifiers
CAS |
629-93-6 |
Molecular Formula |
C18H37I |
Molecular Weight (g/mol) |
380.398 |
MDL Number |
MFCD00001090 |
InChI Key |
ZNJOCVLVYVOUGB-UHFFFAOYSA-N |
Synonym |
Octadecyl Iodide, Stearyl Iodide |
PubChem CID |
12402 |
IUPAC Name |
1-iodooctadecane |
SMILES |
CCCCCCCCCCCCCCCCCCI |
View More Specs
Products
3
Specifications
Chemical Identifiers
|
629-93-6 |
|
380.398 |
|
ZNJOCVLVYVOUGB-UHFFFAOYSA-N |
|
12402 |
|
CCCCCCCCCCCCCCCCCCI |
|
C18H37I |
|
MFCD00001090 |
|
Octadecyl Iodide, Stearyl Iodide |
|
1-iodooctadecane |
Specifications
|
197°C |
|
34°C |
|
629-93-6 |
|
MFCD00001090 |
|
ZNJOCVLVYVOUGB-UHFFFAOYSA-N |
|
1-iodooctadecane |
|
12402 |
|
Crystal at 20°C |
|
1-Iodooctadecane |
|
White-Yellow |
|
25g |
|
C18H37I |
|
Octadecyl Iodide, Stearyl Iodide |
|
CCCCCCCCCCCCCCCCCCI |
|
380.398 |
|
380.40 |
|
≥98.0% (GC) |
Safety and Handling