Please login to your online account to display your discounted pricing

1-Iodononane 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Iodononane
Name Note 99%
CAS 4282-42-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Molecular Formula C9H19I
Linear Formula CH3(CH2)8I
Formula Weight 254.16
Synonym Nonyl iodide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC250070100

  SDS
thermo fisher scientific
AC250070100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC250070500

  SDS
thermo fisher scientific
AC250070500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.16
Physical Form Liquid
Assay ≥98.5%
Quantity 10g
Color Clear colorless to pale yellow
TSCA TSCA
EINECS Number 224-286-3
Density 1.28
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 98.5
Packaging Glass bottle
Stabilizer copper
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Synonym Nonyl iodide
CAS 4282-42-2
Health Hazard 1 LIGHT SENSITIVE
Name Note 99%
Chemical Name or Material 1-Iodononane
Linear Formula CH3(CH2)8I
Molecular Formula C9H19I
Safety and Handling

LIGHT SENSITIVE