Please login to your online account to display your discounted pricing

1-Iodononane 99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC250070500

 View more versions of this product

Catalog No. AC250070500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 254.16
Physical Form Liquid
Assay ≥98.5%
Quantity 50g
Color Clear colorless to pale yellow
TSCA TSCA
EINECS Number 224-286-3
Density 1.28
CAS Max % 100.0
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Health Hazard 1 LIGHT SENSITIVE
CAS Min % 98.5
Packaging Glass bottle
Stabilizer copper
Synonym Nonyl iodide
Infrared Spectrum Authentic
Linear Formula CH3(CH2)8I
Molecular Formula C9H19I
Name Note 99%
CAS 4282-42-2
Chemical Name or Material 1-Iodononane

Specifications

Chemical Name or Material 1-Iodononane
Name Note 99%
CAS 4282-42-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Molecular Formula C9H19I
Linear Formula CH3(CH2)8I
Formula Weight 254.16
Synonym Nonyl iodide
Accessories

LIGHT SENSITIVE

SDS & Safety Documents